Learn More
Sino Biological Human CD28 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11524R00720
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD28 / TP44 (rh CD28 / TP44; Catalog#11524-H08H; P10747-1; Met1-Pro152).
Catalog No. 50-237-3118