Learn More
ACROBiosystems ACROBiosystems Human CD3 delta Protein, His Tag
Catalog No. 502107494
Encompass_Preferred
Recombinant Protein;100UG;Human CD3 delta, His Tag (CDD-H5224) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Ala 105 (Accession # P04234-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.4 kDa. The protein migrates as 18-28 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-210-7494 Supplier ACROBiosystems Supplier No. CDDH5224100UG
Recombinant Protein;100UG;Human CD3 delta, His Tag (CDD-H5224) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Ala 105 (Accession # P04234-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.4 kDa. The protein migrates as 18-28 kDa under reducing (R) condition (SDS-PAGE).