Learn More
ACROBiosystems ACROBiosystems Human CD3 epsilon&CD3 delta Heterodimer Protein, Fc Tag&Fc Tag (MALS verified)
Catalog No. 502106968
Encompass_Preferred
Recombinant Protein;100UG;Human CD3E&CD3D Heterodimer Protein, Fc Tag&Fc Tag (CDD-H5255) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) & P04234-1 (CD3D)).;Human CD3E&CD3D Heterodimer Protein, Fc Tag&Fc Tag is produced by co-expression of CD3E and CD3D, has a calculated MW of 38.3 kDa (CD3E) and 35.8 kDa (CD3D). Subunit CD3E is fused with a human IgG1 Fc tag at the C-terminus and subunit CD3D is fused with a human IgG1 Fc tag at the C-terminus. As a result of glycosylation, the protein migrates as 45-60 kDa under reducing (R) condition, and 90-110 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CD3E&CD3D Heterodimer Protein, Fc Tag&Fc Tag (Cat. No. CDD-H5255) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3), Ultra-low endotoxin (Cat. No. CDE-M120a) with a linear range of 0.8-6 ng/mL (QC tested).;Immobilized Human CD3E&CD3D Heterod
Catalog No. 50-210-6968 Supplier ACROBiosystems Supplier No. CDDH5255100UG
Recombinant Protein;100UG;Human CD3E&CD3D Heterodimer Protein, Fc Tag&Fc Tag (CDD-H5255) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) & P04234-1 (CD3D)).;Human CD3E&CD3D Heterodimer Protein, Fc Tag&Fc Tag is produced by co-expression of CD3E and CD3D, has a calculated MW of 38.3 kDa (CD3E) and 35.8 kDa (CD3D). Subunit CD3E is fused with a human IgG1 Fc tag at the C-terminus and subunit CD3D is fused with a human IgG1 Fc tag at the C-terminus. As a result of glycosylation, the protein migrates as 45-60 kDa under reducing (R) condition, and 90-110 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CD3E&CD3D Heterodimer Protein, Fc Tag&Fc Tag (Cat. No. CDD-H5255) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3), Ultra-low endotoxin (Cat. No. CDE-M120a) with a linear range of 0.8-6 ng/mL (QC tested).;Immobilized Human CD3E&CD3D Heterod