Learn More
ACROBiosystems ACROBiosystems Human CD3 epsilon&CD3 delta Heterodimer Protein, Fc,His Tag&Fc,Flag Tag (MALS verified)
Catalog No. 502107028
Encompass_Preferred
Recombinant Protein;100UG;Human CD3E&CD3D Heterodimer Protein (CDD-H52Wa) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) & P04234-1 (CD3D)).;Human CD3E&CD3D Heterodimer Protein is produced by co-expression of CD3E and CD3D, has a calculated MW of 40.2 kDa (CD3E) and 37.3 kDa (CD3D). The reducing (R) protein migrates as 45-55 kDa due to glycosylation.;Immobilized Human CD3E&CD3D Heterodimer Protein (Cat. No. CDD-H52Wa) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3), Ultra-low endotoxin (Cat. No. CDE-M120a) with a linear range of 0.2-3 ng/mL (QC tested).;Immobilized Human CD3E&CD3D Heterodimer Protein (Cat. No. CDD-H52Wa) at 1 μg/mL (100 μL/well) can bind UCHT1 with a linear range of 0.1-3 ng/mL (Routinely tested).
Catalog No. 50-210-7028 Supplier ACROBiosystems Supplier No. CDDH52WA100UG
Recombinant Protein;100UG;Human CD3E&CD3D Heterodimer Protein (CDD-H52Wa) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) & P04234-1 (CD3D)).;Human CD3E&CD3D Heterodimer Protein is produced by co-expression of CD3E and CD3D, has a calculated MW of 40.2 kDa (CD3E) and 37.3 kDa (CD3D). The reducing (R) protein migrates as 45-55 kDa due to glycosylation.;Immobilized Human CD3E&CD3D Heterodimer Protein (Cat. No. CDD-H52Wa) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3), Ultra-low endotoxin (Cat. No. CDE-M120a) with a linear range of 0.2-3 ng/mL (QC tested).;Immobilized Human CD3E&CD3D Heterodimer Protein (Cat. No. CDD-H52Wa) at 1 μg/mL (100 μL/well) can bind UCHT1 with a linear range of 0.1-3 ng/mL (Routinely tested).