Recombinant Protein;50UG;Human CD3E&CD3G Heterodimer Protein, His Tag&Tag Free (CDG-H52W5) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # P07766-1 (CD3E) & P09693-1 (CD3G)).;Human CD3E&CD3G Heterodimer Protein, His Tag&Tag Free is produced by co-expression of CD3E and CD3G, has a calculated MW of 16.9 kDa (CD3E) and 14.6 kDa (CD3G). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3G contains no tag at the C-terminus. The reducing (R) heterodimer protein migrates as 19 kDa and 21-28 kDa due to glycosylation.;Immobilized Human CD3E&CD3G Heterodimer Protein, His Tag&Tag Free (Cat. No. CDG-H52W5) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3) with a linear range of 0.1-3 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3) at 5 μg/mL (100 μL/well) can bind Human CD3E&CD3G Heterodimer Protein, His T