Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CD3 epsilon Heterodimer Recombinant Protein

Catalog No. PIRP87999 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP87999 500 μg
PIRP87998 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP87999 Supplier Invitrogen™ Supplier No. RP87999
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (200 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.

CD3E and CD3D Heterodimer Protein is produced by mammalian expression system and are involved in mediating signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04234, P07766
Concentration 1.0 mg/mL
For Use With (Application) Control, ELISA Standard, Binding Assay
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose and no preservative; pH 7.4
Gene ID (Entrez) 915, 916
Molecular Weight (g/mol) 77.5 kDa
Name Human CD3 epsilon Heterodimer
pH Range 7.4
Purification Method Purified
Quantity 500 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Common Name CD3e/CD3d
Gene Symbol CD3D, CD3E
Biological Activity Measured by its ability to bind OKT3-mIgG2a in a functional ELISA.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Human CD3D, Met1-Ala105 (Accession # P04234-1) with a C-terminal His-tag hIgG1 Fc region and human CD3E, Met1-Asp126 (Accession # P07766-1) with a C-terminal Flag-tag hIgG Fc region were co-expressed and purified as a human CD3D & CD3E heterodimer
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Immobilized SP34 Antibody at 2 μg/mL (100 μL/well) can bind Recombinant Human CD3D & CD3E Heterodimer Protein (ECD) (Cat: CT026-H0323H) , the EC50 is 8-25 ng/mL.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥90% as determined by SDS-PAGE. ≥95% as determined by SEC-HPLC.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.