Learn More
ACROBiosystems Human CD3 gamma / CD3G Protein, Fc Tag (MALS verified)
Catalog No. 502054758
Encompass_Preferred
Recombinant Protein;1MG;Human CD3 gamma, Fc Tag (CDA-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 116 (Accession # P09693-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 37.1 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4758 Supplier ACROBiosystems Supplier No. CDAH52541MG
Recombinant Protein;1MG;Human CD3 gamma, Fc Tag (CDA-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 116 (Accession # P09693-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 37.1 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.