Learn More
R&D Systems™ Human CD30/TNFRSF8 DuoSet ELISA

Description
Includes
Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Specifications
Specifications
| Assay Range | 93.80 to 6000 pg/mL |
| Detectable Analytes | CD30/TNFRSF8 |
| Assay | Solid Phase Sandwich ELISA |
| Conjugate | Biotin |
| Cross Reactivity | <50%cross-species reactivity observed with species tested |
| Immunoassay Kit Format | 96-well Strip Plate |
| Product Type | ELISA Development Kit |
| Sample Type | Cell Culture Supernatant |
| Sufficient For | To run ELISAs on at least 5 x 96-well plates |
| For Use With (Application) | ELISA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.