Learn More
ACROBiosystems Human CD30 / TNFRSF8 Protein, His Tag
Supplier:  ACROBiosystems CD0H52H51MG
Encompass_Preferred
Recombinant Protein;1MG;Human CD30, His Tag (CD0-H52H5) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.4 kDa.
Catalog No. 50-205-4888