Learn More
Sino Biological Human CD300A MAb 100ul (Sino Biological)
Supplier:  Sino Biological 12449MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD300A (rh CD300A; Catalog#12449-H08H; Q9UGN4-1; Met1-Gln178). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2545