Learn More
Sino Biological Anti-CD300A/LMIR1 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 12449MM01F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD300A (rh CD300A; Catalog#12449-H08H; Q9UGN4-1; Met1-Gln178) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-2499