Learn More
Sino Biological Human CD300A MAb 100TS (Sino Biological)
Supplier:  Sino Biological 12449MM01A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD300A (rh CD300A; Catalog#12449-H08H; Q9UGN4-1; Met1-Gln178) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-3372