Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CD300e/LMIR6 ELISA Kit

Catalog No. EH85RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH85RBX5 5 x 96 Tests
EH85RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH85RBX5 Supplier Invitrogen™ Supplier No. EH85RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human CD300e/LMIR6 quantitates human CD300e/LMIR6 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD300e/LMIR6.

CD300e is a member of the glycoprotein family of cell surface proteins expressed on myeloid cells. CD300e interacts with the TYRO protein tyrosine kinase-binding protein and is thought to be involved as an activating receptor. CD300e is expressed on mature myeloid dendritic cells and monocytes, and is down regulated on immature dendritic cells. Further, CD300e interacts with DAP-12, which mediates activating signals. The alloreactive response of naïve T cells has been reported to become enhanced by the activation of CD300e. Diseases associated with CD300e dysfunction include tungiasis and lice infestation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q496F6
Assay Range 0.5-30 ng/mL
Assay Sensitivity 0.5 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias BC034097; CD300 antigen like family member E; CD300 antigen-like family member E; CD300e; CD300e antigen; CD300e molecule; Cd300le; CLM2; CLM-2; CMRF35A5; CMRF35-A5; CMRF35-like molecule 2; CMRF-35-like molecule-1; Immune receptor expressed on myeloid cells 2; IREM2; IREM-2; PIgR2; PIgR-2; poly-Ig receptor 2; polymeric immunoglobulin receptor 2; TREM5BC034097; CD300 antigen like family member E; CD300 antigen-like family member E; CD300e; CD300e antigen; CD300e molecule; Cd300le; CLM2; CLM-2; CMRF35A5; CMRF35-A5; CMRF35-like molecule 2; CMRF-35-like molecule-1; Immune receptor expressed on myeloid cells 2; IREM2; IREM-2; PIgR2; PIgR-2; poly-Ig receptor 2; polymeric immunoglobulin receptor 2; TREM5;
Gene ID (Entrez) 342510
Gene Symbol CD300E
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target CD300e
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.