Learn More
Raybiotech Inc Human CD300e/LMIR6 ELISA Kit
Supplier:  Raybiotech Inc ELHCD300E1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human CD300e.
Catalog No. 50-194-9081