Learn More
Sino Biological Recombinant Human CD300f/LMIR3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 16034H08H50
Encompass_Preferred
A DNA sequence encoding the human CD300LF (NP_620587.2) (Met1-Leu155) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4070