Learn More
Biotang Inc Human CD31 / PECAM1 Protein (His Tag), 100ug, the extracellular domain (Met 1-Lys 601) of human CD31 (EAW94208.1) was expressed in human cells with a polyhistidine tag at the C-terminus.
Supplier:  Biotang Inc RPR9794
Encompass_Preferred
Human CD31 / PECAM1 Protein (His Tag), 100ug, the extracellular domain (Met 1-Lys 601) of human CD31 (EAW94208.1) was expressed in human cells with a polyhistidine tag at the C-terminus.
Catalog No. 50-751-5408