Learn More
ACROBiosystems Human CD32b/c Protein Strep T
Catalog No. 502535319
Encompass_Preferred
Human CD32b/c Protein Strep Tag (CDB-H5284) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession # P31994-1).
Catalog No. 50-253-5319 Supplier ACROBiosystems Supplier No. CDBH528450UG
Human CD32b/c Protein Strep Tag (CDB-H5284) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession # P31994-1).