Learn More
Sino Biological Human CD32B/Fcgr2b Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785765
Encompass_Preferred
The human CD32b / FCGR2B ELISA Pair Set is for the quantitative determinationof human CD32b / FCGR2B.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD32b / FCGR2B coated on a 96-well plate. Standards and samples are added to the wells, and any CD32b / FCGR2B present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-CD32b / FCGR2B monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD32b / FCGR2B present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85765 Supplier Sino Biological Supplier No. SEKA1025915
The human CD32b / FCGR2B ELISA Pair Set is for the quantitative determinationof human CD32b / FCGR2B.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD32b / FCGR2B coated on a 96-well plate. Standards and samples are added to the wells, and any CD32b / FCGR2B present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-CD32b / FCGR2B monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD32b / FCGR2B present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.