Learn More
Sino Biological Anti-Siglec-3/CD33 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 12238MM06F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD33/Siglec-3 (rh CD33/Siglec-3; Catalog#12238-H08H; AAH28152.1; Met1-His259) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-2059