Learn More
Sino Biological Recombinant Anti-Siglec-3/CD33 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12238R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD33 / Siglec-3 (rh CD33 / Siglec-3; Catalog#12238-H08H; AAH28152.1; Met1-His259).
Catalog No. 50-241-1776