Learn More
Sino Biological Recombinant Human Siglec-3/CD33 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 12238H02H100
Encompass_Preferred
A DNA sequence encoding the human CD33 (AAH28152.1) (Met1-His259) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4397