Learn More
Sino Biological Human CD33 / Siglec-3 Protein (PE conjugated)
Supplier:  Sino Biological 12238HCCHP25
Encompass_Preferred
Recombinant human CD33 (AAH28152.1, Met1-His259) are conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-3100