Learn More
ACROBiosystems Human CD34 200ug
Supplier:  ACROBiosystems CD4H82E3200UG
Encompass_Preferred
Biotinylated Human CD34 Protein HisAvitag (CD4-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Thr 290 (Accession # P28906-1).
Catalog No. 50-253-5379