Learn More
ACROBiosystems Human CD34 25ug
Catalog No. 502535378
Encompass_Preferred
Biotinylated Human CD34 Protein HisAvitag (CD4-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Thr 290 (Accession # P28906-1).
Catalog No. 50-253-5378 Supplier ACROBiosystems Supplier No. CD4H82E325UG
Biotinylated Human CD34 Protein HisAvitag (CD4-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Thr 290 (Accession # P28906-1).