Learn More
Lonza Walkersville Human CD34+ Cell Nucleofector Kit
Catalog No. NC9584304
Encompass
The Nucleofector 2b Device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector Kit is the optimal kit for efficient transfection of primary human hematopoietic stem cells (CD34+) from cord blood or bone marrow.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells Low-throughput transfection in single cuvette formatContent:2.25 mL Nucleofector Solution for Human CD34+ Cells0.5 mL Supplement 130 µg pmaxGFP Vector (0.5 μg/μl in 10 mM Tris pH 8.0)25 Aluminum cuvette (100 µL)25 Single use pipettes
Catalog No. NC9584304 Supplier Lonza Walkersville Supplier No. VPA1003
The Nucleofector 2b Device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector Kit is the optimal kit for efficient transfection of primary human hematopoietic stem cells (CD34+) from cord blood or bone marrow.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells Low-throughput transfection in single cuvette formatContent:2.25 mL Nucleofector Solution for Human CD34+ Cells0.5 mL Supplement 130 µg pmaxGFP Vector (0.5 μg/μl in 10 mM Tris pH 8.0)25 Aluminum cuvette (100 µL)25 Single use pipettes