Learn More
Sino Biological Anti-CD34 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10103MM26100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with full length human CD34 expression plasmid. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1233