Learn More
Lonza Walkersville Human CD34+ Progenitor Cells from Mobilized Peripheral Blood, Positive Selection, 1 Million
Supplier:  Lonza Walkersville 4Y101C
Encompass
Mobilized peripheral blood CD34+ hematopoietic stem and progenitor cells are positively selected using a immunomagnetic CD34 microbead labeling system. Cells are cryopreserved in Lonza Cryo Media in a cryovial. Cell purity is determined post thaw using a FACS assay. Count and viability is determined using AO/PI. Cells are collected from healthy donors following IRB protocols.Manufactured by AllCellsBenefits:Obtain large quantities of CD34+ from a donorCertificate of analysis (CoA) provided for each lot purchasedTest negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsApplications:CAR-T therapy developmentiPSC generationAutoimmune diseaseImmunologyCancer researchContent:1 x Cryopreserved ampule of Human CD34+ Progenitor Cells from Mobilized Peripheral Blood containing ≥ 1 million cells
Catalog No. NC1940515