Learn More
Sino Biological Recombinant Human CD34 Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10103H02HB100
Encompass_Preferred
A DNA sequence encoding the human CD34 precursor (NP_001020280.1) (Met 1-Thr 290) was fused with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5479