Learn More
Sino Biological Human CD34 Protein 20ug (Sino Biological)
Supplier:  Sino Biological 10103H49HB20
Encompass_Preferred
A DNA sequence encoding the Human CD34 (NP_001020280.1) (Met1-Thr290) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-1416