Learn More
Bioworld HUMAN CD34 (RECOMBINANT)
Supplier:  Bioworld 220602801
Encompass_Preferred
Recombinant Human CD34 Source: E.Coli Background: CD34 is a cell surface antigen formerly known as hemopoietic progenitor cell antigen-1. CD34 is expressed selectively on human hematopoietic progenitor cells . The biological function...
Catalog No. 50-253-379