Learn More
ACROBiosystems Human CD36 25ug
Catalog No. 502535360
Encompass_Preferred
Biotinylated Human CD36 Protein FcAvitag (CD6-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # P16671-1).
Catalog No. 50-253-5360 Supplier ACROBiosystems Supplier No. CD6H82F425UG
Biotinylated Human CD36 Protein FcAvitag (CD6-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # P16671-1).