Learn More
Sino Biological Recombinant Human CD36 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10752H49HB100
Encompass_Preferred
A DNA sequence encoding the Human CD36 (NP_001001547.1)(Gly30-Asn439) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8747