Learn More
ACROBiosystems Human CD36 Protein Fc Tag
Supplier:  ACROBiosystems CD6H5258100UG
Encompass_Preferred
Human CD36 Protein Fc Tag (CD6-H5258) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # P16671-1).
Catalog No. 50-253-5365