Learn More
Raybiotech Inc Human CD36/SR-B3 ELISA Kit
Supplier:  Raybiotech Inc ELHCD361
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human CD36.
Catalog No. 50-194-9086