Learn More
Sino Biological Anti-CD38 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10818MM22100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD38 (rh CD38; Catalog#10818-H08H; NP_001766.2; Val43-Ile300). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9538