Learn More
Sino Biological Recombinant Anti-CD38 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10818R067100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD38 / ADPRC1 (rh CD38 / ADPRC1; Catalog#10818-H08H; NP_001766.2; Val43-Ile300).
Catalog No. 50-241-1825