Learn More
Sino Biological Anti-CD38 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10818MM27F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD38/ADPRC1 (rh CD38/ADPRC1; Catalog#10818-H08H; NP_001766.2; Val43-Ile300) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-9638