Learn More
Sino Biological Anti-CD38 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10818T26100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD38 (rh CD38; Catalog#10818-H08H; NP_001766.2; Val43-Ile300). CD38 specific IgG was purified by Human CD38 affinity chromatography.
Catalog No. 50-255-5151