Learn More
Sino Biological Anti-CD38 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10818RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD38 (rhCD38; Catalog#10818-H08H; NP_001766.2; Val 43-Ile 300). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8561