Learn More
Sino Biological Recombinant Human CD38 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10818H27HB100
Encompass_Preferred
A DNA sequence encoding the human CD38 (NP_001766.2) (Val43-Ile300) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5156