Learn More
BIOHIPPO Human CD39 Microsample
SDP
Catalog No. 5000313702
Encompass
Human CD39 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD39 in serum plasma and biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD39 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD39 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD39 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13702 Supplier BIOHIPPO Supplier No. BHE15212355
Human CD39 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD39 in serum plasma and biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD39 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD39 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD39 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color