Learn More
Sino Biological Recombinant Anti-CD3D & CD3E Heterodimer Antibody (PerCP), Rabbit Monoclonal
Supplier:  Sino Biological CT026R301C100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD3D & CD3E (rh CD3D & CD3E; Catalog#CT026-H0323H; P04234-1 & P07766-1; Met1-Ala105 & Met1-Asp126) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-241-1469