Learn More
Sino Biological Human CD3D & CD3E MAb 100TS (Sino Biological)
Supplier:  Sino Biological CT026R301P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD3D & CD3E (rh CD3D & CD3E; Catalog#CT026-H0323H; P04234-1 & P07766-1; Met1-Ala105 & Met1-Asp126) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-1254