Learn More
Sino Biological Recombinant Anti-CD3D & CD3E Heterodimer Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological CT026R301F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD3D & CD3E Heterodimer (Catalog#CT026-H0323H; P04234-1 & P07766-1; Met1-Ala105 & Met1-Asp126) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-7852