Learn More
Sino Biological Anti-CD3 delta/CD3D Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10981MM0820
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD3D (rh CD3D; Catalog#10981-H08H; NP_000723.1; Met 1-Ala 105). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8768