Learn More
BIOHIPPO Human CD3d Microsample
SDP
Catalog No. 5000315105
Encompass
Human CD3d Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD3d in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 112 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD3d Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD3d Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD3d biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15105 Supplier BIOHIPPO Supplier No. BHE15211334
Human CD3d Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD3d in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 112 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD3d Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD3d Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD3d biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color