Learn More
Sino Biological Recombinant Human CD3 delta/CD3D Protein (hFc & FLAG Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10981H23HB100
Encompass_Preferred
A DNA sequence encoding the human CD3D (NP_000723.1) (Met1-Ala105) was fused with the C-terminal flag-tagged Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4997