Learn More
Sino Biological Recombinant Human CD3D & CD3E Heterodimer Protein (Flag & His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological CT038H2508HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Ala105) of human CD3D (P04234) was fused with a flag tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the extracellular domain (Met1-Asp126) of human CD3E (P07766) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the human CD3D/CD3E heterodimer was purified. The purified protein was biotinylated in vitro.
Catalog No. 50-255-2614