Learn More
Sino Biological Recombinant Human CD3E & CD3G Heterodimer Protein (C-Llama-Fc), HPLC-verified
Supplier:  Sino Biological CT089H46HE100
Encompass_Preferred
A DNA sequence encoding the human CD3E (NP_000724.1) (Met1-Asp126) was fused with the Fc region of Llama IgG2b at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the human CD3G (NP_000064.1) (Met1-Ser113) was fused with the Fc region of Llama IgG2b at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the human CD3E/CD3G heterodimer was purified.
Catalog No. 50-255-2601