Learn More
Sino Biological Anti-CD4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10400T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD4 (rh CD4; Catalog#10400-H08H; NP_000607.1; Met1-Trp390). CD4 specific IgG was purified by Human CD4 affinity chromatography.
Catalog No. 50-255-5319